Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MOSPD2
Entrez Gene ID
158747
Gene Name
motile sperm domain containing 2
Gene Aliases
-
Chromosome Location
Chr.X: 14873405 - 14922166 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177475.1 NP_001170946.1 10 - 11 1178 116
NM_152581.3 NP_689794.1 11 - 12 1214 116
XM_005274451.1 XP_005274508.1 11 - 12 1214 116
GenBank mRNA AK093075.1 - 10 - 11 1178 116
AK315781.1 - 11 - 12 1154 115
AL834345.1 - 3 - 4 266 116
BC030641.1 - 11 - 12 1181 116

View Full Product Details