The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161407.1 | NP_001154879.1 | 5 - 6 | 858 | 88 |
NM_019080.2 | NP_061953.2 | 5 - 6 | 918 | 88 | |
XM_006719840.3 | XP_006719903.1 | 5 - 6 | 1317 | 88 | |
GenBank mRNA | AB032991.1 | - | 5 - 6 | 682 | 88 |
AB097019.1 | - | 5 - 6 | 890 | 88 | |
AB097028.1 | - | 5 - 6 | 890 | 88 | |
AB097029.1 | - | 5 - 6 | 890 | 88 | |
AB097030.1 | - | 5 - 6 | 568 | 88 | |
AB097031.1 | - | 5 - 6 | 568 | 88 | |
AK001723.1 | - | 5 - 6 | 709 | 88 | |
AK294839.1 | - | 5 - 6 | 529 | 88 | |
AK297197.1 | - | 5 - 6 | 625 | 88 | |
BC021988.1 | - | 5 - 6 | 650 | 88 | |
BC026126.2 | - | 5 - 6 | 679 | 88 |