The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001089.2 | NP_001080.2 | 9 - 10 | 1700 | 59 |
GenBank mRNA | AB070929.1 | - | 6 - 7 | 1030 | 59 |
AB210044.1 | - | 9 - 10 | 1700 | 59 | |
AK310864.1 | - | 9 - 10 | 1551 | 59 | |
BC140895.1 | - | 6 - 7 | 1025 | 59 | |
BC146866.1 | - | 6 - 7 | 1025 | 59 | |
BC151840.1 | - | 9 - 10 | 1682 | 59 | |
U78735.1 | - | 9 - 10 | 1547 | 59 | |
X97187.1 | - | 8 - 9 | 1335 | 59 |