Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GGN
Entrez Gene ID
199720
Gene Name
gametogenetin
Gene Aliases
-
Chromosome Location
Chr.19: 38384347 - 38388603 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152657.3 NP_689870.3 1 - 2 53 90
GenBank mRNA AF538035.1 - 1 - 2 58 90
AF538036.1 - 1 - 2 58 90
AF538037.1 - 1 - 2 49 90
BC035008.2 - 1 - 2 53 90

View Full Product Details