The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000455.4 | NP_000446.1 | 6 - 7 | 1978 | 73 |
XM_005259617.2 | XP_005259674.1 | 6 - 7 | 1488 | 73 | |
XM_005259618.3 | XP_005259675.1 | 6 - 7 | 1488 | 73 | |
XM_011528209.1 | XP_011526511.1 | 7 - 8 | 1619 | 73 | |
GenBank mRNA | AF035625.1 | - | 6 - 7 | 1201 | 73 |
AF217978.1 | - | 7 - 8 | 1824 | 73 | |
AK314858.1 | - | 6 - 7 | 989 | 73 | |
BC007981.2 | - | 6 - 7 | 1313 | 73 | |
BC019334.1 | - | 6 - 7 | 1134 | 73 | |
EF644172.1 | - | 6 - 7 | 947 | 73 | |
KU178339.1 | - | 6 - 7 | 863 | 73 | |
U63333.1 | - | 6 - 7 | 863 | 73 |