The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014674.2 | NP_055489.1 | 10 - 11 | 1807 | 119 |
XM_011534271.2 | XP_011532573.1 | 10 - 11 | 2001 | 119 | |
XM_011534272.2 | XP_011532574.1 | 11 - 12 | 1800 | 119 | |
GenBank mRNA | AK292643.1 | - | 10 - 11 | 1675 | 119 |
BC019088.1 | - | 10 - 11 | 1814 | 119 | |
D86967.1 | - | 10 - 11 | 1733 | 119 |