The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014674.2 | NP_055489.1 | 11 - 12 | 2015 | 89 |
XM_011534271.2 | XP_011532573.1 | 11 - 12 | 2209 | 89 | |
XM_011534272.2 | XP_011532574.1 | 12 - 13 | 2008 | 89 | |
GenBank mRNA | AK292643.1 | - | 11 - 12 | 1883 | 89 |
BC019088.1 | - | 11 - 12 | 2022 | 89 | |
D86967.1 | - | 11 - 12 | 1941 | 89 |