Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ITPR1
Entrez Gene ID
3708
Gene Name
inositol 1,4,5-trisphosphate receptor type 1
Gene Aliases
ACV, CLA4, INSP3R1, IP3R, IP3R1, PPP1R94, SCA15, SCA16, SCA29
Chromosome Location
Chr.3: 4493348 - 4847840 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099952.2 NP_001093422.2 33 - 34 4599 95
NM_001168272.1 NP_001161744.1 32 - 33 4581 95
NM_002222.5 NP_002213.5 32 - 33 4554 95
XM_005265109.2 XP_005265166.1 33 - 34 4626 95
XM_005265110.2 XP_005265167.1 33 - 34 4626 95
XM_006713131.2 XP_006713194.1 33 - 34 4626 95
XM_011533681.1 XP_011531983.1 33 - 34 4626 95
XM_011533682.2 XP_011531984.1 33 - 34 4552 95
XM_011533683.2 XP_011531985.1 33 - 34 4626 95
XM_011533684.1 XP_011531986.1 33 - 34 4599 95
XM_011533685.1 XP_011531987.1 33 - 34 4626 95
XM_011533686.1 XP_011531988.1 33 - 34 4626 95
XM_011533687.1 XP_011531989.1 32 - 33 4581 95
XM_011533688.1 XP_011531990.1 32 - 33 4554 95
XM_011533690.1 XP_011531992.1 33 - 34 4626 95
XM_011533691.1 XP_011531993.1 33 - 34 4626 95
XM_011533692.2 XP_011531994.1 33 - 34 4626 95
XM_017006357.1 XP_016861846.1 33 - 34 4626 95
XM_017006358.1 XP_016861847.1 33 - 34 4626 95
GenBank mRNA AB208868.1 - 10 - 11 1354 95
AW501800.1 - 2 - 3 95
D26070.1 - 32 - 33 4460 95
L38019.2 - 33 - 34 4496 95
U23850.1 - 30 - 31 4231 95

View Full Product Details