The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099952.2 | NP_001093422.2 | 57 - 58 | 8231 | 86 |
NM_001168272.1 | NP_001161744.1 | 59 - 60 | 8330 | 86 | |
NM_002222.5 | NP_002213.5 | 56 - 57 | 8186 | 86 | |
XM_005265109.2 | XP_005265166.1 | 58 - 59 | 8306 | 86 | |
XM_005265110.2 | XP_005265167.1 | 57 - 58 | 8258 | 86 | |
XM_006713131.2 | XP_006713194.1 | 58 - 59 | 8309 | 86 | |
XM_011533681.1 | XP_011531983.1 | 60 - 61 | 8378 | 86 | |
XM_011533682.2 | XP_011531984.1 | 60 - 61 | 8304 | 86 | |
XM_011533683.2 | XP_011531985.1 | 60 - 61 | 8375 | 86 | |
XM_011533684.1 | XP_011531986.1 | 60 - 61 | 8351 | 86 | |
XM_011533685.1 | XP_011531987.1 | 59 - 60 | 8345 | 86 | |
XM_011533686.1 | XP_011531988.1 | 59 - 60 | 8342 | 86 | |
XM_011533687.1 | XP_011531989.1 | 59 - 60 | 8333 | 86 | |
XM_011533688.1 | XP_011531990.1 | 59 - 60 | 8306 | 86 | |
XM_011533690.1 | XP_011531992.1 | 59 - 60 | 8255 | 86 | |
XM_017006357.1 | XP_016861846.1 | 59 - 60 | 8252 | 86 | |
GenBank mRNA | AB208868.1 | - | 37 - 38 | 5102 | 86 |
AK294594.1 | - | 8 - 9 | 1303 | 86 | |
AK296235.1 | - | 8 - 9 | 1327 | 86 | |
AK309981.1 | - | 8 - 9 | 1282 | 86 | |
BC144563.1 | - | 18 - 19 | 2062 | 86 | |
D26070.1 | - | 56 - 57 | 8092 | 86 | |
L38019.2 | - | 57 - 58 | 8128 | 86 | |
U23850.1 | - | 57 - 58 | 7980 | 86 |