The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002068.3 | NP_002059.3 | 3 - 4 | 901 | 71 |
GenBank mRNA | AF493904.1 | - | 3 - 4 | 482 | 71 |
AK291329.1 | - | 3 - 4 | 691 | 71 | |
BC013585.1 | - | 3 - 4 | 667 | 71 | |
BT009850.1 | - | 3 - 4 | 482 | 71 | |
GU727637.1 | - | 3 - 4 | 667 | 71 | |
GU727645.1 | - | 3 - 4 | 740 | 71 | |
M63904.1 | - | 3 - 4 | 701 | 71 |