The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001285.3 | NP_001276.2 | 10 - 11 | 1814 | 62 |
GenBank mRNA | AF039400.1 | - | 11 - 12 | 2036 | 62 |
AF127036.1 | - | 10 - 11 | 1689 | 62 | |
AK024970.1 | - | 5 - 6 | 898 | 62 | |
AK097057.1 | - | 10 - 11 | 1547 | 62 | |
AK291999.1 | - | 10 - 11 | 1730 | 62 | |
AK300858.1 | - | 6 - 7 | 1100 | 62 | |
AK314375.1 | - | 10 - 11 | 1732 | 62 | |
AW361520.1 | - | 3 - 4 | 533 | 62 |