The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144633.2 | NP_653234.2 | 2 - 3 | 510 | 132 |
XM_017005698.1 | XP_016861187.1 | 2 - 3 | 581 | 132 | |
XM_017005700.1 | XP_016861189.1 | 2 - 3 | 581 | 132 | |
XM_017005701.1 | XP_016861190.1 | 2 - 3 | 568 | 132 | |
GenBank mRNA | AB209053.1 | - | 2 - 3 | 501 | 132 |
AK295633.1 | - | 2 - 3 | 557 | 132 | |
AY053503.1 | - | 2 - 3 | 510 | 132 |