Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STXBP4
Entrez Gene ID
252983
Gene Name
syntaxin binding protein 4
Gene Aliases
Synip
Chromosome Location
Chr.17: 54968728 - 55164285 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178509.5 NP_848604.3 11 - 12 1141 96
XM_005257187.4 XP_005257244.1 11 - 12 1183 96
XM_017024410.1 XP_016879899.1 12 - 13 1232 96
XM_017024411.1 XP_016879900.1 12 - 13 1224 96
XM_017024412.1 XP_016879901.1 11 - 12 1175 96
XM_017024413.1 XP_016879902.1 9 - 10 896 96
GenBank mRNA AK122865.1 - 11 - 12 1141 96

View Full Product Details