Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STXBP4
Entrez Gene ID
252983
Gene Name
syntaxin binding protein 4
Gene Aliases
Synip
Chromosome Location
Chr.17: 54968728 - 55164285 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178509.5 NP_848604.3 7 - 8 784 100
XM_005257187.4 XP_005257244.1 7 - 8 826 100
XM_006721797.3 XP_006721860.1 7 - 8 824 100
XM_006721798.3 XP_006721861.1 7 - 8 815 100
XM_017024410.1 XP_016879899.1 8 - 9 875 100
XM_017024411.1 XP_016879900.1 8 - 9 867 100
XM_017024412.1 XP_016879901.1 7 - 8 818 100
XM_017024413.1 XP_016879902.1 5 - 6 539 100
XM_017024414.1 XP_016879903.1 8 - 9 874 100
XM_017024415.1 XP_016879904.1 8 - 9 874 100
GenBank mRNA AK122865.1 - 7 - 8 784 100
BC041485.2 - 6 - 7 627 100
BC125116.1 - 7 - 8 773 100

View Full Product Details