The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153710.4 | NP_714921.4 | 5 - 6 | 458 | 76 |
GenBank mRNA | BC036504.2 | - | 5 - 6 | 458 | 76 |