Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADAM11
Entrez Gene ID
4185
Gene Name
ADAM metallopeptidase domain 11
Gene Aliases
MDC
Chromosome Location
Chr.17: 44759037 - 44781846 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318933.1 NP_001305862.1 17 - 18 1503 102
NM_002390.5 NP_002381.2 17 - 18 1507 102
XM_005257373.3 XP_005257430.1 17 - 18 1649 102
XM_017024647.1 XP_016880136.1 16 - 17 1421 102
GenBank mRNA AB009675.1 - 17 - 18 1486 102
AK296512.1 - 17 - 18 1584 102
CD630842.1 - 3 - 4 102
CN418442.1 - 1 - 2 120 102
D17390.1 - 14 - 15 1216 102

View Full Product Details