Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADAM11
Entrez Gene ID
4185
Gene Name
ADAM metallopeptidase domain 11
Gene Aliases
MDC
Chromosome Location
Chr.17: 44759037 - 44781846 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318933.1 NP_001305862.1 23 - 24 2092 84
NM_002390.5 NP_002381.2 23 - 24 2096 84
XM_005257373.3 XP_005257430.1 23 - 24 2238 84
XM_017024647.1 XP_016880136.1 22 - 23 2010 84
GenBank mRNA AB009675.1 - 23 - 24 2075 84
AK296512.1 - 23 - 24 2173 84
D17390.1 - 20 - 21 1815 84

View Full Product Details