Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADAM11
Entrez Gene ID
4185
Gene Name
ADAM metallopeptidase domain 11
Gene Aliases
MDC
Chromosome Location
Chr.17: 44759037 - 44781846 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002390.5 NP_002381.2 8 - 9 699 64
XM_005257373.3 XP_005257430.1 8 - 9 841 64
GenBank mRNA AB009675.1 - 8 - 9 678 64
CD630831.1 - 6 - 7 64
D17390.1 - 5 - 6 408 64

View Full Product Details