Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRC2
Entrez Gene ID
9902
Gene Name
mannose receptor C type 2
Gene Aliases
CD280, CLEC13E, ENDO180, UPARAP
Chromosome Location
Chr.17: 62627401 - 62693601 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006039.4 NP_006030.2 26 - 27 4299 93
GenBank mRNA AB014609.2 - 26 - 27 4299 93
AF107292.1 - 26 - 27 3940 93
AF134838.1 - 26 - 27 4021 93
AK093230.1 - 7 - 8 1514 93
BC146647.1 - 26 - 27 4018 93
BC150212.1 - 26 - 27 4003 93
BC153884.1 - 26 - 27 4299 93
U58856.1 - 2 - 3 245 93

View Full Product Details