The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006039.4 | NP_006030.2 | 8 - 9 | 1860 | 94 |
XM_011525543.2 | XP_011523845.1 | 8 - 9 | 1591 | 94 | |
GenBank mRNA | AB014609.2 | - | 8 - 9 | 1860 | 94 |
AF107292.1 | - | 8 - 9 | 1501 | 94 | |
AF134838.1 | - | 8 - 9 | 1582 | 94 | |
BC146647.1 | - | 8 - 9 | 1579 | 94 | |
BC150212.1 | - | 8 - 9 | 1564 | 94 | |
BC153884.1 | - | 8 - 9 | 1860 | 94 |