Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRC2
Entrez Gene ID
9902
Gene Name
mannose receptor C type 2
Gene Aliases
CD280, CLEC13E, ENDO180, UPARAP
Chromosome Location
Chr.17: 62627401 - 62693601 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006039.4 NP_006030.2 9 - 10 1968 118
XM_011525543.2 XP_011523845.1 9 - 10 1699 118
GenBank mRNA AB014609.2 - 9 - 10 1968 118
AF107292.1 - 9 - 10 1609 118
AF134838.1 - 9 - 10 1690 118
BC146647.1 - 9 - 10 1687 118
BC150212.1 - 9 - 10 1672 118
BC153884.1 - 9 - 10 1968 118

View Full Product Details