The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005264779.4 | XP_005264836.2 | 1 - 2 | 428 | 70 |
XM_017005490.1 | XP_016860979.1 | 1 - 2 | 420 | 70 | |
GenBank mRNA | BG818516.1 | - | 1 - 2 | 365 | 70 |