The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003931.3 | NP_001003931.3 | 2 - 3 | 587 | 64 |
NM_005485.5 | NP_005476.4 | 2 - 3 | 592 | 64 | |
XM_005264779.4 | XP_005264836.2 | 3 - 4 | 732 | 64 | |
XM_017005490.1 | XP_016860979.1 | 3 - 4 | 719 | 64 | |
GenBank mRNA | AB208803.1 | - | 2 - 3 | 472 | 64 |
AF083068.1 | - | 2 - 3 | 311 | 64 | |
AY126341.1 | - | 2 - 3 | 422 | 64 | |
BC014260.2 | - | 1 - 2 | 193 | 64 | |
BI554046.1 | - | 2 - 3 | 526 | 64 | |
Y16836.1 | - | 2 - 3 | 546 | 64 |