The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001305792.1 | NP_001292721.1 | 6 - 7 | 1773 | 108 |
NM_014875.2 | NP_055690.1 | 8 - 9 | 2185 | 108 | |
XM_011510231.2 | XP_011508533.1 | 9 - 10 | 2279 | 108 | |
XM_011510232.2 | XP_011508534.1 | 8 - 9 | 1941 | 108 | |
XM_011510233.2 | XP_011508535.1 | 9 - 10 | 2072 | 108 | |
XM_011510235.2 | XP_011508537.1 | 9 - 10 | 1784 | 108 | |
XM_011510236.1 | XP_011508538.1 | 7 - 8 | 2068 | 108 | |
XM_017003005.1 | XP_016858494.1 | 7 - 8 | 2362 | 108 | |
XM_017003006.1 | XP_016858495.1 | 8 - 9 | 1970 | 108 | |
XM_017003007.1 | XP_016858496.1 | 8 - 9 | 1497 | 108 | |
GenBank mRNA | BC113742.1 | - | 7 - 8 | 1824 | 108 |
D26361.1 | - | 8 - 9 | 2185 | 108 |