The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114978.1 | NP_001108450.1 | 9 - 10 | 1308 | 90 |
NM_001114979.1 | NP_001108451.1 | 9 - 10 | 1308 | 90 | |
NM_001114980.1 | NP_001108452.1 | 7 - 8 | 1078 | 90 | |
NM_001114981.1 | NP_001108453.1 | 7 - 8 | 1078 | 90 | |
NM_001114982.1 | NP_001108454.1 | 7 - 8 | 1078 | 90 | |
NM_003722.4 | NP_003713.3 | 9 - 10 | 1308 | 90 | |
XM_005247843.3 | XP_005247900.1 | 9 - 10 | 1743 | 90 | |
XM_005247844.4 | XP_005247901.1 | 9 - 10 | 1471 | 90 | |
XM_011513251.2 | XP_011511553.1 | 10 - 11 | 1349 | 90 | |
XM_011513252.2 | XP_011511554.1 | 9 - 10 | 1281 | 90 | |
XM_017007387.1 | XP_016862876.1 | 7 - 8 | 1074 | 90 | |
GenBank mRNA | AB010153.1 | - | 7 - 8 | 995 | 90 |
AB016072.1 | - | 9 - 10 | 1246 | 90 | |
AB016073.1 | - | 9 - 10 | 1246 | 90 | |
AB042841.1 | - | 6 - 7 | 682 | 90 | |
AF075428.1 | - | 8 - 9 | 1102 | 90 | |
AF075429.1 | - | 7 - 8 | 937 | 90 | |
AF075430.1 | - | 8 - 9 | 1102 | 90 | |
AF075431.1 | - | 7 - 8 | 937 | 90 | |
AF075432.1 | - | 8 - 9 | 1102 | 90 | |
AF075433.1 | - | 7 - 8 | 937 | 90 | |
AF091627.1 | - | 7 - 8 | 1062 | 90 | |
AF116771.1 | - | 9 - 10 | 1246 | 90 | |
AK304127.1 | - | 7 - 8 | 1078 | 90 | |
BC039815.1 | - | 9 - 10 | 1308 | 90 | |
GQ202689.1 | - | 7 - 8 | 1076 | 90 | |
GQ202690.1 | - | 7 - 8 | 1076 | 90 | |
KP881229.1 | - | 7 - 8 | 942 | 90 | |
KP881230.1 | - | 7 - 8 | 930 | 90 | |
KP881231.1 | - | 6 - 7 | 675 | 90 | |
KU178435.1 | - | 9 - 10 | 1219 | 90 | |
KU178436.1 | - | 9 - 10 | 1207 | 90 | |
Y16961.1 | - | 9 - 10 | 1246 | 90 |