Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ASTN1
Entrez Gene ID
460
Gene Name
astrotactin 1
Gene Aliases
ASTN
Chromosome Location
Chr.1: 176855118 - 177164904 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004319.2 NP_004310.1 23 - 23 153
XM_017001340.1 XP_016856829.1 23 - 23 153
GenBank mRNA AB006627.1 - 23 - 23 153
BC146795.1 - 23 - 23 153
BU620695.1 - 1 - 1 140 153

View Full Product Details