The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142610.1 | NP_001136082.1 | 1 - 2 | 602 | 112 |
NM_014683.3 | NP_055498.3 | 1 - 2 | 602 | 112 | |
XM_017025424.1 | XP_016880913.1 | 1 - 2 | 427 | 112 | |
XM_017025425.1 | XP_016880914.1 | 1 - 2 | 427 | 112 | |
XM_017025426.1 | XP_016880915.1 | 1 - 2 | 427 | 112 | |
GenBank mRNA | AB014523.1 | - | 1 - 2 | 593 | 112 |
AK314345.1 | - | 1 - 2 | 127 | 112 | |
BC034988.1 | - | 2 - 3 | 167 | 112 |