The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142610.1 | NP_001136082.1 | 7 - 8 | 1049 | 89 |
NM_014683.3 | NP_055498.3 | 7 - 8 | 1049 | 89 | |
XM_011524087.2 | XP_011522389.1 | 3 - 4 | 346 | 89 | |
XM_017025424.1 | XP_016880913.1 | 7 - 8 | 937 | 89 | |
XM_017025425.1 | XP_016880914.1 | 7 - 8 | 937 | 89 | |
XM_017025426.1 | XP_016880915.1 | 7 - 8 | 937 | 89 | |
XM_017025427.1 | XP_016880916.1 | 6 - 7 | 450 | 89 | |
XM_017025428.1 | XP_016880917.1 | 6 - 7 | 450 | 89 | |
GenBank mRNA | AB014523.1 | - | 7 - 8 | 1040 | 89 |
AK098316.1 | - | 6 - 7 | 674 | 89 | |
AK314345.1 | - | 7 - 8 | 574 | 89 | |
BC034988.1 | - | 8 - 9 | 614 | 89 |