Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OFCC1
Entrez Gene ID
266553
Gene Name
orofacial cleft 1 candidate 1
Gene Aliases
MRDS1
Chromosome Location
Chr.6: 9775808 - 9842630 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF520803.1 - 4 - 5 376 89
AF520805.1 - 8 - 9 817 89
AF520806.1 - 7 - 8 657 89
AF548114.1 - - 387 89
AF548115.1 - 4 - 5 442 89
AF548116.1 - 5 - 6 528 89
AF548122.1 - 3 - 4 411 89
AF548123.1 - 5 - 6 481 89

View Full Product Details