The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004326.3 | NP_004317.2 | 6 - 7 | 1207 | 67 |
XM_005272971.4 | XP_005273028.1 | 6 - 7 | 1212 | 67 | |
XM_005277417.3 | XP_005277474.1 | 6 - 7 | 1236 | 67 | |
XM_005277418.3 | XP_005277475.1 | 6 - 7 | 1092 | 67 | |
XM_017002026.1 | XP_016857515.1 | 6 - 7 | 990 | 67 | |
GenBank mRNA | BC116451.1 | - | 3 - 4 | 602 | 67 |
Y13620.1 | - | 6 - 7 | 1295 | 67 |