Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYBL1
Entrez Gene ID
4603
Gene Name
MYB proto-oncogene like 1
Gene Aliases
A-MYB, AMYB
Chromosome Location
Chr.8: 66562175 - 66613290 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080416.3 NP_001073885.1 3 - 4 605 83
NM_001144755.2 NP_001138227.1 3 - 4 605 83
NM_001294282.1 NP_001281211.1 3 - 4 605 83
XM_011517533.2 XP_011515835.1 3 - 4 646 83
XM_011517535.2 XP_011515837.1 3 - 4 646 83
XM_017013455.1 XP_016868944.1 3 - 4 456 83
XM_017013456.1 XP_016868945.1 3 - 4 453 83
XM_017013457.1 XP_016868946.1 3 - 4 454 83
XM_017013458.1 XP_016868947.1 3 - 4 451 83
XM_017013459.1 XP_016868948.1 3 - 4 458 83
GenBank mRNA BC101186.1 - 3 - 4 263 83
BC101188.1 - 3 - 4 263 83
X13269.1 - 3 - 4 298 83
X13294.1 - 3 - 4 298 83

View Full Product Details