Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NOL11
Entrez Gene ID
25926
Gene Name
nucleolar protein 11
Gene Aliases
-
Chromosome Location
Chr.17: 67717833 - 67744208 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001303272.1 NP_001290201.1 12 - 13 1586 134
NM_015462.4 NP_056277.2 13 - 14 1644 134
GenBank mRNA AK023702.1 - 13 - 14 1534 134
AK025390.1 - 2 - 3 572 134
AK299483.1 - 12 - 13 1476 134
AL110271.1 - 10 - 11 1234 134
AY598333.1 - 13 - 14 1529 134
BC001726.2 - 5 - 6 598 134
BC064404.1 - 13 - 14 1535 134

View Full Product Details