Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NOL11
Entrez Gene ID
25926
Gene Name
nucleolar protein 11
Gene Aliases
-
Chromosome Location
Chr.17: 67717833 - 67744208 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001303272.1 NP_001290201.1 2 - 3 365 165
NM_015462.4 NP_056277.2 2 - 3 365 165
GenBank mRNA AK023702.1 - 2 - 3 255 165
AK299483.1 - 2 - 3 255 165
AY598333.1 - 2 - 3 250 165
BC064404.1 - 2 - 3 256 165
DA772356.1 - 2 - 3 365 165

View Full Product Details