The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286229.1 | NP_001273158.1 | 4 - 5 | 679 | 76 |
NM_001286230.1 | NP_001273159.1 | 4 - 5 | 679 | 76 | |
NM_016448.3 | NP_057532.3 | 5 - 6 | 805 | 76 | |
XM_011509614.1 | XP_011507916.1 | 5 - 6 | 509 | 76 | |
GenBank mRNA | AF195765.1 | - | 5 - 6 | 569 | 76 |
AF345896.1 | - | 5 - 6 | 590 | 76 | |
AK000742.1 | - | 5 - 6 | 623 | 76 | |
AK292343.1 | - | 5 - 6 | 677 | 76 | |
AK303346.1 | - | 4 - 5 | 482 | 76 | |
BC033297.1 | - | 5 - 6 | 546 | 76 | |
BC033540.1 | - | 5 - 6 | 587 | 76 | |
DQ641253.1 | - | 5 - 6 | 467 | 76 |