The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000965.4 | NP_000956.2 | 1 - 1 | 527 | 91 |
NM_001290276.1 | NP_001277205.1 | 1 - 1 | 527 | 91 | |
NM_001290277.1 | NP_001277206.1 | 1 - 1 | 527 | 91 | |
NR_110892.1 | 1 - 1 | 527 | 91 | ||
NR_110893.1 | 1 - 1 | 527 | 91 | ||
GenBank mRNA | AB307688.1 | - | 1 - 1 | 58 | 91 |
AK292032.1 | - | 1 - 1 | 77 | 91 | |
BC060794.1 | - | 1 - 1 | 209 | 91 | |
CD014020.1 | - | 1 - 1 | 74 | 91 | |
CD014021.1 | - | 1 - 1 | 74 | 91 | |
CD014022.1 | - | 1 - 1 | 74 | 91 | |
HQ692864.1 | - | 1 - 1 | 58 | 91 | |
X07282.1 | - | 1 - 1 | 401 | 91 | |
Y00291.1 | - | 1 - 1 | 379 | 91 |