The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000249.3 | NP_000240.1 | 5 - 6 | 654 | 80 |
NM_001167617.1 | NP_001161089.1 | 5 - 6 | 449 | 80 | |
NM_001167618.1 | NP_001161090.1 | 5 - 6 | 449 | 80 | |
NM_001258271.1 | NP_001245200.1 | 5 - 6 | 654 | 80 | |
NM_001258273.1 | NP_001245202.1 | 4 - 5 | 563 | 80 | |
NM_001258274.1 | NP_001245203.1 | 6 - 7 | 599 | 80 | |
XM_005265161.1 | XP_005265218.1 | 5 - 6 | 544 | 80 | |
XM_005265163.1 | XP_005265220.1 | 5 - 6 | 454 | 80 | |
XM_005265164.1 | XP_005265221.1 | 5 - 6 | 507 | 80 | |
XM_005265166.1 | XP_005265223.1 | 5 - 6 | 462 | 80 | |
XM_011533727.1 | XP_011532029.1 | 5 - 6 | 462 | 80 | |
XM_017006449.1 | XP_016861938.1 | 5 - 6 | 462 | 80 | |
XM_017006450.1 | XP_016861939.1 | 5 - 6 | 507 | 80 | |
XM_017006451.1 | XP_016861940.1 | 5 - 6 | 507 | 80 | |
GenBank mRNA | AB209848.1 | - | 5 - 6 | 431 | 81 |
AK222810.1 | - | 5 - 6 | 461 | 80 | |
AK295359.1 | - | 5 - 6 | 428 | 80 | |
AK298324.1 | - | 5 - 6 | 399 | 81 | |
AK298583.1 | - | 5 - 6 | 422 | 80 | |
AK302807.1 | - | 7 - 8 | 383 | 80 | |
AK311326.1 | - | 5 - 6 | 664 | 80 | |
AK312609.1 | - | 5 - 6 | 470 | 80 | |
AK316264.1 | - | 4 - 5 | 380 | 80 | |
AY517558.1 | - | 5 - 6 | 454 | 80 | |
BC006850.1 | - | 5 - 6 | 506 | 80 | |
BX648844.1 | - | 6 - 7 | 602 | 81 | |
DA517219.1 | - | 5 - 6 | 454 | 80 | |
U07343.1 | - | 5 - 6 | 477 | 80 | |
U07418.1 | - | 5 - 6 | 497 | 80 |