The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013436.4 | NP_038464.1 | 15 - 16 | 2237 | 168 |
NM_205842.2 | NP_995314.1 | 16 - 17 | 2255 | 168 | |
XM_006712200.3 | XP_006712263.1 | 16 - 17 | 1741 | 168 | |
XM_006712201.3 | XP_006712264.1 | 15 - 16 | 1725 | 168 | |
GenBank mRNA | AB011159.1 | - | 16 - 17 | 2255 | 168 |
AB014509.1 | - | 15 - 16 | 1497 | 168 | |
AK292914.1 | - | 15 - 16 | 1727 | 168 | |
BC015025.1 | - | 15 - 16 | 1749 | 168 | |
BT007033.1 | - | 15 - 16 | 1478 | 168 |