The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013436.4 | NP_038464.1 | 20 - 21 | 2907 | 107 |
NM_205842.2 | NP_995314.1 | 21 - 22 | 2925 | 107 | |
XM_006712200.3 | XP_006712263.1 | 21 - 22 | 2411 | 107 | |
XM_006712201.3 | XP_006712264.1 | 20 - 21 | 2395 | 107 | |
GenBank mRNA | AB011159.1 | - | 21 - 22 | 2925 | 107 |
AB014509.1 | - | 20 - 21 | 2167 | 107 | |
AK001291.1 | - | 1 - 2 | 118 | 107 | |
AK002153.2 | - | 3 - 4 | 403 | 107 | |
AK292914.1 | - | 20 - 21 | 2397 | 107 | |
BC015025.1 | - | 20 - 21 | 2419 | 107 | |
BT007033.1 | - | 20 - 21 | 2148 | 107 |