The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042663.1 | NP_001036128.1 | 21 - 22 | 3278 | 83 |
NM_001042664.1 | NP_001036129.1 | 20 - 21 | 3232 | 83 | |
NM_001042665.1 | NP_001036130.1 | 20 - 21 | 3215 | 83 | |
NM_001265592.1 | NP_001252521.1 | 21 - 22 | 3311 | 83 | |
NM_001265593.1 | NP_001252522.1 | 20 - 21 | 3242 | 83 | |
NM_001265594.1 | NP_001252523.1 | 21 - 22 | 3046 | 83 | |
NM_020631.4 | NP_065682.2 | 20 - 21 | 3161 | 83 | |
NM_198681.3 | NP_941374.2 | 21 - 22 | 3790 | 83 | |
GenBank mRNA | AB018263.2 | - | 21 - 22 | 3266 | 83 |
AB097001.1 | - | 19 - 20 | 3001 | 83 | |
AK024676.1 | - | 20 - 21 | 3160 | 83 | |
AK091201.1 | - | 8 - 9 | 2211 | 83 | |
AK096347.1 | - | 21 - 22 | 3738 | 83 | |
AK125550.1 | - | 21 - 22 | 3244 | 83 | |
AK131074.1 | - | 18 - 19 | 3485 | 83 | |
AK294875.1 | - | 21 - 22 | 3311 | 83 | |
AK299523.1 | - | 20 - 21 | 3242 | 83 | |
AK301239.1 | - | 14 - 15 | 2241 | 83 | |
BC015231.1 | - | 21 - 22 | 3022 | 83 | |
BC036671.2 | - | 21 - 22 | 3098 | 83 |