The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042663.1 | NP_001036128.1 | 5 - 6 | 479 | 95 |
NM_001042664.1 | NP_001036129.1 | 4 - 5 | 433 | 95 | |
NM_001042665.1 | NP_001036130.1 | 4 - 5 | 416 | 95 | |
NM_001265592.1 | NP_001252521.1 | 5 - 6 | 512 | 95 | |
NM_001265593.1 | NP_001252522.1 | 4 - 5 | 443 | 95 | |
NM_001265594.1 | NP_001252523.1 | 4 - 5 | 447 | 95 | |
NM_020631.4 | NP_065682.2 | 4 - 5 | 362 | 95 | |
NM_198681.3 | NP_941374.2 | 5 - 6 | 991 | 95 | |
GenBank mRNA | AB018263.2 | - | 5 - 6 | 467 | 95 |
AB097001.1 | - | 3 - 4 | 208 | 95 | |
AK024676.1 | - | 4 - 5 | 361 | 95 | |
AK096347.1 | - | 5 - 6 | 939 | 95 | |
AK125550.1 | - | 5 - 6 | 445 | 95 | |
AK131074.1 | - | 3 - 4 | 227 | 95 | |
AK294875.1 | - | 5 - 6 | 512 | 95 | |
AK299523.1 | - | 4 - 5 | 443 | 95 | |
BC015231.1 | - | 4 - 5 | 423 | 95 | |
BC036671.2 | - | 5 - 6 | 992 | 95 | |
DA091930.1 | - | 4 - 5 | 447 | 95 | |
DA100291.1 | - | 4 - 5 | 433 | 95 | |
DA126163.1 | - | 4 - 5 | 416 | 95 | |
DB093234.1 | - | 4 - 5 | 362 | 95 | |
DB225012.1 | - | 5 - 6 | 479 | 95 |