The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042663.1 | NP_001036128.1 | 10 - 11 | 1254 | 135 |
NM_001042664.1 | NP_001036129.1 | 9 - 10 | 1208 | 135 | |
NM_001042665.1 | NP_001036130.1 | 9 - 10 | 1191 | 135 | |
NM_001265592.1 | NP_001252521.1 | 10 - 11 | 1287 | 135 | |
NM_001265593.1 | NP_001252522.1 | 9 - 10 | 1218 | 135 | |
NM_001265594.1 | NP_001252523.1 | 9 - 10 | 1222 | 135 | |
NM_020631.4 | NP_065682.2 | 9 - 10 | 1137 | 135 | |
NM_198681.3 | NP_941374.2 | 10 - 11 | 1766 | 135 | |
GenBank mRNA | AB018263.2 | - | 10 - 11 | 1242 | 135 |
AK024676.1 | - | 9 - 10 | 1136 | 135 | |
AK096347.1 | - | 10 - 11 | 1714 | 135 | |
AK125550.1 | - | 10 - 11 | 1220 | 135 | |
AK131074.1 | - | 7 - 8 | 1461 | 135 | |
AK294875.1 | - | 10 - 11 | 1287 | 135 | |
AK299523.1 | - | 9 - 10 | 1218 | 135 | |
AK301239.1 | - | 3 - 4 | 217 | 135 | |
BC015231.1 | - | 9 - 10 | 1198 | 135 | |
BC036671.2 | - | 10 - 11 | 1767 | 135 |