Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TANC1
Entrez Gene ID
85461
Gene Name
tetratricopeptide repeat, ankyrin repeat and coiled-coil containing 1
Gene Aliases
ROLSB, TANC
Chromosome Location
Chr.2: 158968600 - 159232659 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145909.1 NP_001139381.1 11 - 12 1751 67
NM_033394.2 NP_203752.2 11 - 12 1775 67
XM_005246918.4 XP_005246975.1 10 - 11 1539 67
XM_006712810.3 XP_006712873.1 10 - 11 1662 67
XM_006712812.3 XP_006712875.1 10 - 11 1663 67
XM_006712814.3 XP_006712877.1 7 - 8 958 67
XM_011512048.2 XP_011510350.1 13 - 14 1953 67
XM_011512051.2 XP_011510353.1 12 - 13 1818 67
XM_011512053.2 XP_011510355.1 11 - 12 1708 67
XM_011512055.2 XP_011510357.1 11 - 12 1744 67
XM_011512058.2 XP_011510360.1 9 - 10 1450 67
XM_011512059.2 XP_011510361.1 7 - 8 1311 67
XM_011512062.2 XP_011510364.1 5 - 6 988 67
XM_017005141.1 XP_016860630.1 10 - 11 1567 67
XM_017005142.1 XP_016860631.1 12 - 13 2172 67
XM_017005143.1 XP_016860632.1 10 - 11 1857 67
XM_017005144.1 XP_016860633.1 10 - 11 1546 67
XM_017005145.1 XP_016860634.1 13 - 14 1987 67
XM_017005146.1 XP_016860635.1 12 - 13 1798 67
XM_017005147.1 XP_016860636.1 11 - 12 1747 67
XM_017005148.1 XP_016860637.1 12 - 13 1913 67
XM_017005149.1 XP_016860638.1 9 - 10 1465 67
XM_017005150.1 XP_016860639.1 9 - 10 1447 67
XM_017005151.1 XP_016860640.1 9 - 10 1306 67
XM_017005152.1 XP_016860641.1 9 - 10 1303 67
XM_017005153.1 XP_016860642.1 8 - 9 1252 67
XM_017005154.1 XP_016860643.1 11 - 12 1669 67
XM_017005155.1 XP_016860644.1 9 - 10 1399 67
XM_017005156.1 XP_016860645.1 7 - 8 988 67
XM_017005157.1 XP_016860646.1 8 - 9 1165 67
XM_017005158.1 XP_016860647.1 13 - 14 1916 67
GenBank mRNA AB051515.1 - 5 - 6 852 67
AK308899.1 - 6 - 7 918 67
BC037329.1 - 9 - 10 1456 67
BC150597.1 - 10 - 11 1592 67
BX648327.1 - 6 - 7 1035 67

View Full Product Details