Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TANC1
Entrez Gene ID
85461
Gene Name
tetratricopeptide repeat, ankyrin repeat and coiled-coil containing 1
Gene Aliases
ROLSB, TANC
Chromosome Location
Chr.2: 158968600 - 159232659 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145909.1 NP_001139381.1 24 - 25 4144 99
NM_033394.2 NP_203752.2 24 - 25 4168 99
XM_005246918.4 XP_005246975.1 23 - 24 3932 99
XM_006712810.3 XP_006712873.1 23 - 24 4055 99
XM_006712812.3 XP_006712875.1 23 - 24 4056 99
XM_006712814.3 XP_006712877.1 20 - 21 3351 99
XM_011512048.2 XP_011510350.1 26 - 27 4346 99
XM_011512051.2 XP_011510353.1 25 - 26 4211 99
XM_011512053.2 XP_011510355.1 24 - 25 4101 99
XM_011512055.2 XP_011510357.1 24 - 25 4137 99
XM_011512058.2 XP_011510360.1 22 - 23 3843 99
XM_011512059.2 XP_011510361.1 20 - 21 3704 99
XM_011512062.2 XP_011510364.1 18 - 19 3381 99
XM_017005141.1 XP_016860630.1 23 - 24 3960 99
XM_017005142.1 XP_016860631.1 25 - 26 4565 99
XM_017005143.1 XP_016860632.1 23 - 24 4250 99
XM_017005144.1 XP_016860633.1 23 - 24 3939 99
XM_017005145.1 XP_016860634.1 26 - 27 4380 99
XM_017005146.1 XP_016860635.1 25 - 26 4191 99
XM_017005147.1 XP_016860636.1 24 - 25 4140 99
XM_017005148.1 XP_016860637.1 25 - 26 4306 99
XM_017005149.1 XP_016860638.1 22 - 23 3858 99
XM_017005150.1 XP_016860639.1 22 - 23 3840 99
XM_017005151.1 XP_016860640.1 22 - 23 3699 99
XM_017005152.1 XP_016860641.1 22 - 23 3696 99
XM_017005153.1 XP_016860642.1 21 - 22 3645 99
XM_017005154.1 XP_016860643.1 24 - 25 4062 99
XM_017005155.1 XP_016860644.1 22 - 23 3792 99
XM_017005156.1 XP_016860645.1 20 - 21 3381 99
XM_017005157.1 XP_016860646.1 21 - 22 3558 99
GenBank mRNA AB051515.1 - 18 - 19 3245 99
AK128859.1 - 13 - 14 2051 99
AK308899.1 - 19 - 20 3311 99
BC150597.1 - 23 - 24 3985 99
BX648327.1 - 19 - 20 3428 99

View Full Product Details