The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017694.3 | NP_060164.3 | 6 - 7 | 2216 | 88 |
XM_005246654.2 | XP_005246711.1 | 5 - 6 | 2094 | 88 | |
XM_005246656.3 | XP_005246713.1 | 5 - 6 | 2094 | 88 | |
XM_011511368.1 | XP_011509670.1 | 7 - 8 | 2282 | 88 | |
XM_017004392.1 | XP_016859881.1 | 6 - 7 | 2216 | 88 | |
GenBank mRNA | AB472299.1 | - | 4 - 5 | 1892 | 88 |
AK000167.1 | - | 4 - 5 | 994 | 88 | |
AK098085.1 | - | 4 - 5 | 1626 | 88 | |
AK127176.1 | - | 6 - 7 | 2216 | 88 | |
AK225240.1 | - | 4 - 5 | 994 | 88 | |
AK302647.1 | - | 4 - 5 | 494 | 88 | |
AK302753.1 | - | 4 - 5 | 517 | 88 | |
BC050537.2 | - | 4 - 5 | 1155 | 88 |