The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289003.1 | NP_001275932.1 | 15 - 16 | 2916 | 92 |
NM_015020.3 | NP_055835.2 | 16 - 17 | 2770 | 92 | |
GenBank mRNA | AB023148.2 | - | 15 - 16 | 2916 | 92 |
AK001854.1 | - | 4 - 5 | 479 | 92 | |
BC035267.1 | - | 11 - 12 | 1605 | 92 | |
BC129927.1 | - | 15 - 16 | 2397 | 92 | |
BX648157.1 | - | 10 - 11 | 1506 | 92 |