The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001023587.2 | NP_001018881.1 | 4 - 5 | 632 | 68 |
NM_001320032.1 | NP_001306961.1 | 4 - 5 | 632 | 68 | |
NM_005688.3 | NP_005679.2 | 4 - 5 | 632 | 68 | |
NR_135125.1 | 4 - 5 | 632 | 68 | ||
XM_005247058.4 | XP_005247115.1 | 4 - 5 | 2200 | 68 | |
XM_005247059.4 | XP_005247116.1 | 4 - 5 | 622 | 68 | |
XM_011512314.1 | XP_011510616.1 | 4 - 5 | 523 | 68 | |
XM_011512315.1 | XP_011510617.1 | 4 - 5 | 682 | 68 | |
XM_017005493.1 | XP_016860982.1 | 4 - 5 | 682 | 68 | |
XM_017005494.1 | XP_016860983.1 | 4 - 5 | 609 | 68 | |
GenBank mRNA | AB019002.1 | - | 3 - 4 | 498 | 68 |
AB209103.1 | - | 1 - 2 | 142 | 68 | |
AB209454.1 | - | 4 - 5 | 559 | 68 | |
AF104942.1 | - | 4 - 5 | 571 | 68 | |
AF146074.1 | - | 4 - 5 | 642 | 68 | |
AL524917.3 | - | 4 - 5 | 544 | 68 | |
AY196484.1 | - | 3 - 4 | 458 | 68 | |
AY754874.1 | - | 4 - 5 | 571 | 68 | |
AY754875.1 | - | 4 - 5 | 571 | 68 | |
AY754876.1 | - | 4 - 5 | 571 | 68 | |
BC050744.1 | - | 3 - 4 | 510 | 68 | |
BC140771.1 | - | 3 - 4 | 508 | 68 | |
BC142670.1 | - | 4 - 5 | 535 | 68 | |
BC142719.1 | - | 4 - 5 | 545 | 68 | |
U83661.2 | - | 4 - 5 | 565 | 68 |