Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SDCCAG3
Entrez Gene ID
10807
Gene Name
serologically defined colon cancer antigen 3
Gene Aliases
NY-CO-3
Chromosome Location
Chr.9: 136401922 - 136410602 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039707.1 NP_001034796.1 8 - 9 1273 70
NM_001039708.1 NP_001034797.1 6 - 7 1054 70
NM_006643.3 NP_006634.3 7 - 8 1204 70
XM_005266050.3 XP_005266107.1 8 - 9 1200 70
XM_005266051.3 XP_005266108.1 7 - 8 1118 70
XM_011518157.2 XP_011516459.1 7 - 8 1057 70
XM_017014218.1 XP_016869707.1 7 - 8 1374 70
XM_017014219.1 XP_016869708.1 7 - 8 1347 70
GenBank mRNA AF039688.1 - 4 - 5 341 70
AK001296.1 - 6 - 7 833 70
AY129014.1 - 1 - 2 1147 70
AY349357.1 - 7 - 8 894 70
BC014515.1 - 7 - 8 1071 70

View Full Product Details