The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004766.2 | NP_004757.1 | 16 - 17 | 2122 | 104 |
NR_023350.1 | 16 - 17 | 2201 | 104 | ||
XM_017007512.1 | XP_016863001.1 | 17 - 18 | 2307 | 104 | |
XM_017007513.1 | XP_016863002.1 | 15 - 16 | 1868 | 104 | |
GenBank mRNA | AK302943.1 | - | 16 - 17 | 2035 | 104 |
BC000326.2 | - | 16 - 17 | 2068 | 104 | |
X70476.1 | - | 16 - 17 | 2060 | 104 |