Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYLK
Entrez Gene ID
4638
Gene Name
myosin light chain kinase
Gene Aliases
AAT7, KRP, MLCK, MLCK1, MLCK108, MLCK210, MSTP083, MYLK1, smMLCK
Chromosome Location
Chr.3: 123612296 - 123884302 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321309.1 NP_001308238.1 29 - 30 5195 118
NM_053025.3 NP_444253.3 30 - 31 5403 118
NM_053026.3 NP_444254.3 29 - 30 5196 118
XM_011512860.2 XP_011511162.1 27 - 28 5124 118
XM_011512861.2 XP_011511163.1 26 - 27 4920 118
XM_017006469.1 XP_016861958.1 13 - 14 2682 118
XM_017006470.1 XP_016861959.1 12 - 13 1670 118
XM_017006471.1 XP_016861960.1 12 - 13 1720 118
GenBank mRNA AB037663.1 - 13 - 14 2479 118
AF069601.2 - 27 - 28 5033 118
AF069604.1 - 7 - 8 1059 118
AK300610.1 - 12 - 13 1653 118
AY339601.1 - 27 - 28 5121 118
AY424269.1 - 26 - 27 4916 118
AY424270.1 - 27 - 28 5123 118
U48959.2 - 28 - 29 5240 118
X85337.1 - 14 - 15 118
X90870.1 - 5 - 6 756 118

View Full Product Details