The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320728.1 | NP_001307657.1 | 2 - 3 | 395 | 110 |
NM_024610.5 | NP_078886.2 | 2 - 3 | 387 | 110 | |
XM_005247762.2 | XP_005247819.1 | 2 - 3 | 387 | 110 | |
XM_011513130.2 | XP_011511432.1 | 3 - 4 | 305 | 110 | |
GenBank mRNA | AF400663.1 | - | 2 - 3 | 354 | 110 |
AK096705.1 | - | 2 - 3 | 282 | 110 | |
AK289410.1 | - | 2 - 3 | 387 | 110 | |
BC011897.2 | - | 2 - 3 | 371 | 110 | |
BC063629.1 | - | 2 - 3 | 327 | 110 | |
BI912690.1 | - | 2 - 3 | 367 | 110 | |
HY020808.1 | - | 2 - 3 | 398 | 110 |